Skip to Content
Merck
All Photos(2)

Key Documents

SAB3501019

Sigma-Aldrich

Anti-VNN3 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 55 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VNN3(55350)

Immunogen

Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human VNN3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501019.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Granjeaud et al.
Immunogenetics, 49(11-12), 964-972 (1999-09-29)
Circulation and tissue colonization are essential properties of lymphoid cells and involve major families of adhesion molecules (e.g. , integrin, selectin, mucin-like, and molecules from the immunoglobulin superfamily). The mouse Vanin-1 molecule was recently identified and found to be involved
Patrick A M Jansen et al.
The Journal of investigative dermatology, 129(9), 2167-2174 (2009-03-27)
The vanin gene family encodes secreted and membrane-bound ectoenzymes that convert pantetheine into pantothenic acid and cysteamine. Recent studies in a mouse colitis model indicated that vanin-1 has proinflammatory activity and suggest that pantetheinases are potential therapeutic targets in inflammatory
Takeaki Nitto et al.
Gene, 426(1-2), 57-64 (2008-09-23)
Pantetheinase (EC 3.5.1.92) is an enzyme that hydrolyzes pantetheine, an intermediate metabolite of coenzyme A, into pantothenic acid (vitamin B(5)) and cysteamine, a potent antioxidant. The pantetheinase gene family consists of three independent genes, pantetheinase/vanin-1/VNN1, GPI-80/VNN2 and vanin-3/VNN3 that are
F Martin et al.
Immunogenetics, 53(4), 296-306 (2001-08-09)
The mouse Vanin-1 molecule plays a role in thymic reconstitution following damage by irradiation. We recently demonstrated that it is a membrane pantetheinase (EC 3.56.1.-). This molecule is the prototypic member of a larger Vanin family encoded by at least

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service