Skip to Content
Merck
All Photos(1)

Key Documents

SAB2107393

Sigma-Aldrich

Anti-NPTX1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

47 kDa

species reactivity

rat, horse, human, bovine, guinea pig, dog, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NPTX1(4884)
mouse ... Nptx1(18164)

Immunogen

The immunogen for anti-NPTX1 antibody: synthetic peptide derected towards the middle region of human NPTX1

Sequence

Synthetic peptide located within the following region: EGKGGPKNDTEERAKIESALTSLHQRISELEKGQKDNRPGDKFQLTFPLR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongfei Yan et al.
Cancer management and research, 11, 3237-3251 (2019-05-03)
This study aimed to investigate the effect of NPTX1 on the prognosis of gastric cancer (GC), as well as the metastatic process in GC. The Cancer Genome Atlas (TCGA) and Gene Expression Omnibus (GEO) databases were used to analyze the
Xiaofeng Peng et al.
Cell biology international, 42(5), 589-597 (2018-01-19)
Colon cancer is the third most common malignancy and one of the leading causes of cancer-associated mortality worldwide. Neuronal pentraxin 1 (NPTX1) is associated with tumor progression in some types of tumors. However, its expression and role in colon cancer
A D Sabag et al.
Immunologic research, 66(6), 649-654 (2019-01-08)
The presence of T regulatory cells (Tregs) is highly required in normal skin in order to maintain immune tolerance to commensal microbes and to prevent the development of immune-mediated inflammation. Psoriasis is a chronic inflammatory skin disease in which effector
Shun Lu et al.
Genes and immunity, 21(1), 4-12 (2019-05-24)
It has been reported that MicroRNAs (miRNAs) play pivotal roles in the occurrence and progression of a variety of cancers. As reported, miR-4295 promotes cell growth and metastasis in a lot of cancers. Nonetheless, the role and molecular mechanism of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service