Skip to Content
Merck
All Photos(2)

Key Documents

SAB2100807

Sigma-Aldrich

Anti-FDXR antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ADXR, Anti-Ferredoxin reductase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FDXR(2232)

Immunogen

Synthetic peptide directed towards the middle region of human FDXR

Biochem/physiol Actions

FDXR is a mitochondrial flavoprotein that initiates electron transport for cytochromes P450 receiving electrons from NADPH.This gene encodes a mitochondrial flavoprotein that initiates electron transport for cytochromes P450 receiving electrons from NADPH. Multiple alternatively spliced transcript variants of this gene have been described although the full-length nature of only two that encode different isoforms have been determined.

Sequence

Synthetic peptide located within the following region: LDPVDFLGLQDKIKEVPRPRKRLTELLLRTATEKPGPAEAARQASASRAW

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda M Dairiki Shortliffe et al.
The Journal of urology, 191(6), 1913-1919 (2014-02-13)
Testosterone affects male development, maturation and aging but limited data exist on testosterone effects on the juvenile genitourinary system. We hypothesized that testosterone has bladder and kidney developmental effects, and investigated this in juvenile male rats. To examine the testosterone
Naoki Terada et al.
Journal of cellular biochemistry, 115(9), 1505-1515 (2014-03-08)
Prostate cancer is a heterogeneous disease and thus, it is important to understand whether among the heterogeneous collection of cell types, androgen-deprivation insensitive cells exist prior to hormonal manipulation. We established several LNCaP subclones with distinct insensitivities to androgen deprivation
Hung-Ming Lam et al.
Endocrine-related cancer, 21(6), 903-914 (2014-10-08)
Castration-resistant prostate cancer (CRPC) is an advanced-stage prostate cancer (PC) associated with high mortality. We reported that G-1, a selective agonist of G protein-coupled receptor 30 (GPR30), inhibited PC cell growth by inducing G2 cell cycle arrest and arrested PC-3
Tanya Aggarwal et al.
Neurobiology of aging, 35(8), 1929-1938 (2014-03-19)
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disease characterized by selective loss of upper and lower motor neurons and skeletal muscle atrophy. Epidemiologic and experimental evidence suggest the involvement of androgens in ALS pathogenesis, but the mechanism through which
F Tian et al.
Andrologia, 46(8), 859-866 (2013-10-16)
AR3, a major one of androgen receptor (AR) splice variants, has been shown to play a pivotal role in concert with AR signalling in prostate cancer. The present study was undertaken to characterise the expression pattern of AR3 in normal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service