SAB1410966
Anti-ACP2 antibody produced in rabbit
purified immunoglobulin, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 48.3 kDa
species reactivity
human
technique(s)
western blot: 1 μg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ACP2(53)
General description
This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene. (provided by RefSeq)
Immunogen
ACP2 (AAH03160.1, 1 a.a. ~ 423 a.a) full-length human protein.
Sequence
MAGKRSGWSRAALLQLLLGVNLVVMPPTQARSLRFVTLLYRHGDRSPVKTYPKDPYQEEEWPQGFGQLTKEGMLQHWELGQALRQRYHGFLNTSYHRQEVYVRSTDFDRTLMSAEANLAGLFPPNGMQRFNPNISWQPIPVHTVPITEDRLLKFPLGPCPRYEQLQNETRQTPEYQNESSRNAQFLDMVANETGLTDLTLETVWNVYDTLFCEQTHGLRLPPWASPQTMQRLSRLKDFSFRFLFGIYQQAEKARLQGGVLLAQIRKNLTLMATTSQLPKLLVYSAHDTTLVALQMALDVYNGEQAPYASCHIFELYQEDSGNFSVEMYFRNESDKAPWPLSLPGCPHRCPLQDFLRLTEPVVPKDWQQECQLASGPADTEVIVALAVCGSILFLLIVLLLTVLFRMQAQPPGYRHVADGEDHA
Sequence
MAGKRSGWSRAALLQLLLGVNLVVMPPTQARSLRFVTLLYRHGDRSPVKTYPKDPYQEEEWPQGFGQLTKEGMLQHWELGQALRQRYHGFLNTSYHRQEVYVRSTDFDRTLMSAEANLAGLFPPNGMQRFNPNISWQPIPVHTVPITEDRLLKFPLGPCPRYEQLQNETRQTPEYQNESSRNAQFLDMVANETGLTDLTLETVWNVYDTLFCEQTHGLRLPPWASPQTMQRLSRLKDFSFRFLFGIYQQAEKARLQGGVLLAQIRKNLTLMATTSQLPKLLVYSAHDTTLVALQMALDVYNGEQAPYASCHIFELYQEDSGNFSVEMYFRNESDKAPWPLSLPGCPHRCPLQDFLRLTEPVVPKDWQQECQLASGPADTEVIVALAVCGSILFLLIVLLLTVLFRMQAQPPGYRHVADGEDHA
Physical form
Solution in phosphate buffered saline, pH 7.4
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service