Skip to Content
Merck
All Photos(2)

Key Documents

SAB1408176

Sigma-Aldrich

Anti-CREB3L1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

OASIS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~57 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CREB3L1(90993)

Immunogen

CREB3L1 (NP_443086.1, 1 a.a. ~ 519 a.a) full-length human protein.

Sequence
MDAVLEPFPADRLFPGSSFLDLGDLNESDFLNNAHFPEHLDHFTENMEDFSNDLFSSFFDDPVLDEKSPLLDMELDSPTPGIQAEHSYSLSGDSAPQSPLVPIKMEDTTQDAEHGAWALGHKLCSIMVKQEQSPELPVDPLAAPSAMAAAAAMATTPLLGLSPLSRLPIPHQAPGEMTQLPVIKAEPLEVNQFLKVTPEDLVQMPPTPPSSHGSDSDGSQSPRSLPPSSPVRPMARSSTAISTSPLLTAPHKLQGTSGPLLLTEEEKRTLIAEGYPIPTKLPLTKAEEKALKRVRRKIKNKISAQESRRKKKEYVECLEKKVETFTSENNELWKKVETLENANRTLLQQLQKLQTLVTNKISRPYKMAATQTGTCLMVAALCFVLVLGSLVPCLPEFSSGSQTVKEDPLAADGVYTASQMPSRSLLFYDDGAGLWEDGRSTLLPMEPPDGWEINPGGPAEQRPRDHLQHDHLDSTHETTKYLSEAWPKDGGNGTSPDFSHSKEWFHDRDLGPNTTIKLS

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

cAMP-Response Element-Binding 3-Like Protein 1 (CREB3L1) is Required for Decidualization and its Expression is Decreased in Women with Endometriosis.
Ahn JI, et. al.
Current Molecular Medicine, 16, 276-276 (2016)
Epigenetic silencing of CREB3L1 by DNA methylation is associated with high-grade metastatic breast cancers with poor prognosis and is prevalent in triple negative breast cancers.
Ward AK, et. al.
Breast Cancer Research, 18, 12-12 (2016)
J I Ahn et al.
Current molecular medicine, 16(3), 276-287 (2016-02-27)
Endometriosis is a major cause of infertility and pelvic pain, affecting more than 10% of reproductive-aged women. Progesterone resistance has been observed in the endometrium of women with this disease, as evidenced by alterations in progesterone-responsive gene and protein expression.
Alison K Ward et al.
Breast cancer research : BCR, 18(1), 12-12 (2016-01-27)
CREB3L1 (cAMP-responsive element-binding protein 3-like protein 1), a member of the unfolded protein response, has recently been identified as a metastasis suppressor in both breast and bladder cancer. Quantitative real time PCR (qPCR) and immunoblotting were used to determine the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service