SAB1401645
Anti-BMP10 antibody produced in mouse
purified immunoglobulin, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
western blot: 1 μg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... BMP10(27302)
General description
The protein encoded by this gene is a member of the TGF-beta family of growth factors. Data suggest that the similar protein in mouse plays an important role in trabeculation of the embryonic heart. In human, this protein may signal through receptor serine/threonine kinases. (provided by RefSeq)
Immunogen
BMP10 (NP_055297.1, 1 a.a. ~ 424 a.a) full-length human protein.
Sequence
MGSLVLTLCALFCLAAYLVSGSPIMNLEQSPLEEDMSLFGDVFSEQDGVDFNTLLQSMKDEFLKTLNLSDIPTQDSAKVDPPEYMLELYNKFATDRTSMPSANIIRSFKNEDLFSQPVSFNGLRKYPLLFNVSIPHHEEVIMAELRLYTLVQRDRMIYDGVDRKITIFEVLESKGDNEGERNMLVLVSGEIYGTNSEWETFDVTDAIRRWQKSGSSTHQLEVHIESKHDEAEDASSGRLEIDTSAQNKHNPLLIVFSDDQSSDKERKEELNEMISHEQLPELDNLGLDSFSSGPGEEALLQMRSNIIYDSTARIRRNAKGNYCKRTPLYIDFKEIGWDSWIIAPPGYEAYECRGVCNYPLAEHLTPTKHAIIQALVHLKNSQKASKACCVPTKLEPISILYLDKGVVTYKFKYEGMAVSECGCR
Sequence
MGSLVLTLCALFCLAAYLVSGSPIMNLEQSPLEEDMSLFGDVFSEQDGVDFNTLLQSMKDEFLKTLNLSDIPTQDSAKVDPPEYMLELYNKFATDRTSMPSANIIRSFKNEDLFSQPVSFNGLRKYPLLFNVSIPHHEEVIMAELRLYTLVQRDRMIYDGVDRKITIFEVLESKGDNEGERNMLVLVSGEIYGTNSEWETFDVTDAIRRWQKSGSSTHQLEVHIESKHDEAEDASSGRLEIDTSAQNKHNPLLIVFSDDQSSDKERKEELNEMISHEQLPELDNLGLDSFSSGPGEEALLQMRSNIIYDSTARIRRNAKGNYCKRTPLYIDFKEIGWDSWIIAPPGYEAYECRGVCNYPLAEHLTPTKHAIIQALVHLKNSQKASKACCVPTKLEPISILYLDKGVVTYKFKYEGMAVSECGCR
Physical form
Solution in phosphate buffered saline, pH 7.4
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lijun Sun et al.
Journal of cellular biochemistry, 115(11), 1868-1876 (2014-06-07)
Heart disease is among the leading causes of death worldwide, and the limited proliferation of mammalian cardiomyocytes prevents heart regeneration in response to injury. Bone morphogenetic protein-10 (BMP10) exerts multiple roles in various developmental events; however, the effect of BMP10
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service