Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400655

Sigma-Aldrich

Anti-VISA antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CARDIF, Anti-DKFZp666M015, Anti-FLJ27482, Anti-FLJ41962, Anti-IPS-1, Anti-KIAA1271, Anti-MAVS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VISA(57506)

General description

Double-stranded RNA viruses are recognized in a cell type-dependent manner by the transmembrane receptor TLR3 (MIM 603029) or by the cytoplasmic RNA helicases MDA5 (MIM 606951) and RIGI (ROBO3; MIM 608630). These interactions initiate signaling pathways that differ in their initial steps but converge in the activation of the protein kinases IKKA (CHUK; MIM 600664) and IKKB (IKBKB; MIM 603258), which activate NFKB (see MIM 164011), or TBK1 (MIM 604834) and IKKE (IKBKE; MIM 605048), which activate IRF3 (MIM 603734). Activated IRF3 and NFKB induce transcription of IFNB (IFNB1; MIM 147640). For the TLR3 pathway, the intermediary molecule before the pathways converge is the cytoplasmic protein TRIF (TICAM1; MIM 607601). For RIGI, the intermediary protein is mitochondria-bound IPS1 (Sen and Sarkar, 2005 [PubMed 16239922]).[supplied by OMIM

Immunogen

VISA (AAH44952.1, 1 a.a. ~ 540 a.a) full-length human protein.

Sequence
MPFAEDKTYKYICRNFSNFCNVDVVEILPYLPCLTARDQDRLRATCTLSGNRDTLWHLFNTLQRRPGWVEYFIAALRGCELVDLADEVASVYESYQPRTSDRPPDPLEPPSLPAERPGPPTPAAAHSIPYNSCREKEPSYPMPVQETQAPESPGENSEQALQTLSPRAIPRNPDGGPLESSSDLAALSPLTSSGHQEKDTELGSTHTAGATSSLTPSRGPVSPSVSFQPLARSTPRASRLPGPTGSVVSTGTSFSSSSPGLASAGAAEGKQGAESDQAEPIICSSGAEAPANSLPSKVPTTLMPVNTVALKVPANPASVSTVPSKLPTSSKPPGAVPSNALTNPAPSKLPINSTRAGMVPSKVPTSMVLTKVSASTVPTDGSSRNEETPAAPTPAGATGGSSAWLDSSFENRGLGSELSKPGVLASQVDSPFSGCFEDLAISASTSLGMGPCHGPEENEYKSEGTFGIHVAENPSIQLLEGNPGPPADPDGGPRPQADRKFQEREVPCHRPSPGALWLQVAVTGVLVVTLLVVLYRRRLH

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Liu et al.
Cell communication and signaling : CCS, 20(1), 103-103 (2022-07-13)
Cytosolic RNA sensing can elicit immune responses against viral pathogens. However, antiviral responses must be tightly regulated to avoid the uncontrolled production of type I interferons (IFN) that might have deleterious effects on the host. Upon bacterial infection, the germinal
Adnan Y Chowdhury et al.
Virology, 456-457, 300-309 (2014-06-04)
We previously found that human pegivirus (HPgV; formerly GBV-C) NS3 protease activity inhibits Human Immunodeficiency Virus (HIV) replication in a CD4+ T cell line. Given the protease׳s similarity to the Hepatitis C virus (HCV) NS3 protease, we characterized HPgV protease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service