SAB1400586
Anti-AHI1 antibody produced in mouse
IgG fraction of antiserum, buffered aqueous solution
Synonym(s):
Anti-DKFZp686J1653, Anti-FLJ14023, Anti-FLJ20069, Anti-JBTS3, Anti-ORF1, Anti-dJ71N10.1
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
western blot: 1 μg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... AHI1(54806)
General description
Immunogen
Sequence
MPTAESEAKVKTKVRFEELLKTHSDLMREKKKLKKKLVRSEENISPDTIRSNLHYMKETTSDDPDTIRSNLPHIKETTSDDVSAANTNNLKKSTRVTKNKLRNTQLATENPNGDASVEEDKQGKPNKKVIKTVPQLTTQDLKPETPENKVDSTHQKTHTKPQPGVDHQKSEKANEGREETDLEEDEELMQAYQCHVTEEMAKEIKRKIRKKLKEQLTYFPSDTLFHDDKLSSEKRKKKKEVPVFSKAETSTLTISGDTVEGEQKKESSVRSVSSDSHQDDEISSMEQSTEDSMQDDTKPKPKKTKKKTKAVADNNEDVDGDGVHEITSRDSPVYPKCLLDDDLVLGVYIHRTDRLKSDFMISHPMVKIHVVDEHTGQYVKKDDSGRPVSSYYEKENVDYILPIMTQPYDFKQLKSRLPEWEEQIVFNENFPYLLRGSDESPKVILFFEILDFLSVDEIKNNSEVQNQECGFRKIAWAFLKLLGANGNANINSKLRLQLYYPPTKPRSPLSVVEAFEWWSKCPRNHYPSTLYVTVRGLKVPDCIKPSYRSMMALQEEKGKPVHCERHHESSSVDTEPGLEESKEVIKWKRLPGQENEDVFVLISPTMEEY
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service