Skip to Content
Merck
All Photos(2)

Key Documents

PLA0061

Sigma-Aldrich

Rabbit anti-MCM3 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

DNA polymerase alpha holoenzyme-associated protein P1, DNA replication factor MCM3, HCC5, MCM3 minichromosome maintenance deficient 3, MCM3 minichromosome maintenance deficient 3 (S. cerevisiae), P1-MCM3, P1.h, RLF subunit beta, RLFB, beta subunit, cervical cancer proto-oncogene 5, hRlf beta subunit, minichromosome maintenance deficient (S. cerevisiae) 3, minichromosome maintenance deficient 3, p102, replication licensing factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunohistochemistry: 1:1,000-1:5,000
immunoprecipitation (IP): 2- 5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_002379.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... MCM3(4172)

Immunogen

The epitope recognized by PLA0061 maps to a region between residues 500 and 550 of human Minichromosome Maintenance 3 using the numbering given in entry NP_002379.2 (GeneID 4172).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

The MCM (Mini-Chromosome Maintenance) complex is a key component of the pre-replication complex involved in replication licensing which restricts DNA replication to only once per cell cycle. The MCM complex is a heterohexamer of MCM2, MCM3, MCM4, MCM5, MCM6, and MCM7. MCM3 has been shown to interact with MCM5, CDC46, and the acetyltransferase MCM3AP.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyle P Heim et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(44), 15746-15751 (2014-10-22)
The cariogenic bacterium Streptococcus mutans uses adhesin P1 to adhere to tooth surfaces, extracellular matrix components, and other bacteria. A composite model of P1 based on partial crystal structures revealed an unusual complex architecture in which the protein forms an
Tatiana Pochechueva et al.
Journal of immunological methods, 412, 42-52 (2014-07-06)
Glycan-based suspension array (SGA) is an "in-house" developed multi-target immunoassay, employing commercially available fluorescent microbeads as a solid support for unique chemically synthesized glycopolymers which capture naturally occurring human anti-glycan antibodies. SGA is a sensitive and reliable tool for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service