P5247
Anti-Potassium Channel K2P9.1 (TASK3) antibody produced in rabbit
affinity isolated antibody, lyophilized powder
Synonym(s):
Anti-Acid Sensitive Potassium Channel 3, Anti-KCNK9
About This Item
Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
rat, human
technique(s)
western blot: 1:200 using rat cerebellum lysate
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... KCNK9(51305)
rat ... Kcnk9(84429)
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service