MTCC2715
MTC™ Bio SpectraTube™ Centrifuge Tubes
capacity 15 mL, assorted colors, pack of 500 ea (10 x racks 50 ea, 5 colours per rack)
Synonym(s):
centrifuge tubes, conical tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Recommended Products
material
assorted colors
clear
conical bottom
polypropylene
screw cap
sterility
sterile
feature
graduation
packaging
pack of 500 ea (10 x racks 50 ea, 5 colours per rack)
manufacturer/tradename
MTC™ Bio C2715
parameter
12,000 × g speed
capacity
15 mL
diam. × L
17 mm × 121 mm
suitability
suitable for (stem cell research)
Related Categories
General description
SpectraTubes Rainbow Capped Centrifuge Tubes, 15mL, sterile, 5 colors per rack (B,G,R,Y,O), 10 racks of 50 tubes, 500/cs
These conical 15mL and 50mL centrifuge tubes are made of premium polypropylene, with exceptionally uniform wall thickness for clarity and centrifugal strength. Tubes and caps may be used for sample storage at temperatures as low as -86°C and are suitable for stem cell research and certified sterile, RNase, DNase, and Pyrogen free.
Features and Benefits
- Color coded caps for easy sample identification
- 15mL rated to 12,000xg
- 50mL rated to 15,000xg
- Sterile, RNase, DNase, pyrogen and cytotoxin free
- Sterile (gamma) and individually packed
- Leak Proof
Legal Information
MTC is a trademark of MTC Bio
SpectraTube is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service