Skip to Content
Merck
All Photos(4)

Key Documents

HPA029909

Sigma-Aldrich

Anti-XPO5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1

Synonym(s):

Anti-KIAA1291, Anti-PRPF10, Anti-Prp10, Anti-SAP155, Anti-SF3b155, Anti-exportin 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL

immunogen sequence

MSFCVYSILGVVKRTCWPTDLEEAKAGGFVVGYTSSGNPIFRNPCTEQILKLLDNLLALIRTHNTLYAPEMLAKMAEPFTKALDMLDAEKSAILGLPQPLLELNDSPVFKTVLERMQRFFSTLYENC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XPO5(57510)

General description

XPO5 (exportin 5) is a Ran-GTP-dependent nuclear transport protein for double-stranded RNA binding proteins. It belongs to the karyopherin protein family which is involved in nucleus to cytoplasm transport of specific classes of small RNAs.

Immunogen

exportin 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

XPO5 (exportin 5) is responsible for the translocation of miRNA (micro-RNA) precursor from nucleus to cytoplasm. Studies in HeLa cells show that miR-138 lowers the stability of XPO5 protein, thus, decreasing miRNA processing. This protein is also involved in the transport of short hairpin (shRNA). It docks on the nuclear pore complex and transports three classes of RNAs, tRNA, Y-RNA and miRNA precursors, across the nuclear membrane, in a RanGTP-RanGDP gradient-dependent manner.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73958

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exportin-5 mediates nuclear export of minihelix-containing RNAs.
Gwizdek C et al
The Journal of Biological Chemistry, 278(8), 5505-5508 (2003)
Overexpression of exportin 5 enhances RNA interference mediated by short hairpin RNAs and microRNAs.
Yi R et al
RNA, 11(2), 220-226 (2005)
Yamina Bennasser et al.
Nature structural & molecular biology, 18(3), 323-327 (2011-02-08)
MicroRNAs (miRNAs) are a class of small, noncoding RNAs that function by regulating gene expression post-transcriptionally. Alterations in miRNA expression can strongly influence cellular physiology. Here we demonstrated cross-regulation between two components of the RNA interference (RNAi) machinery in human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service