Skip to Content
Merck
All Photos(2)

Key Documents

HPA021030

Sigma-Aldrich

Anti-CPA4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Carboxypeptidase A3, Anti-Carboxypeptidase A4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

QVLRINVRNGDEISKLSQLVNSNNLKLNFWKSPSSFNRPVDVLVPSVSLQAFKSFLRSQGLEYAVTIEDLQALLDNEDDEMQHNEGQERSSNNFNYGAYHSLEAIYHEMDNIAADFP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CPA4(51200)

General description

Carboxypeptidase A4 (CPA4), a zinc-containing exopeptidase, is secreted from a soluble proenzyme (pro-CPA4). It is a member of the family of Zn-containing metallocarboxypeptidases with a characteristic α/β-hydrolase fold. The CPA4 gene is mapped to human chromosome 7q32.2.

Immunogen

Carboxypeptidase A4 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CPA4 antibody produced in rabbit has been used in immunohistochemistry and immunostaining (1:300).

Biochem/physiol Actions

Carboxypeptidase A4 (CPA4) catalyzes the release of carboxy-terminal amino acids. It regulates inflammation. An elevated expression of CPA4 is observed in esophageal, pancreatic, and lung tumors. It is implicated in prostate cancer aggressiveness.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74964

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lichao Sun et al.
Journal of Cancer, 8(1), 111-116 (2017-01-27)
The development of liver cancer would undergo a sequential progression from chronic inflammatory liver disease, cirrhosis to neoplasia. During these pathophysiological changes, abnormal liver microenvironment might induce the hepatocytes to die, abnormally proliferate and initiate cancer stem cells. Metallocarboxypeptidases (MCPs)
Lichao Sun et al.
American journal of translational research, 8(11), 5071-5075 (2016-12-03)
CarboxypeptidaseA4 (CPA4) is a zinc-containing exopeptidases, and its aberrant expression has been implicated in cancer development and progression. However, few studies have investigated the association between CPA4 over-expression and clinical significance in gastric cancer (GC). In this study, we employed
Tadashi Handa et al.
International journal of oncology, 54(3), 833-844 (2019-01-11)
Using whole transcriptome analysis and a lentiviral short hairpin RNA screening library, carboxypeptidase A4 (CPA4) was identified as a novel marker in breast cancer and a therapeutic target in triple‑negative breast cancer (TNBC) in the present study. Immunohistochemistry was used to evaluate the
Lichao Sun et al.
American journal of cancer research, 6(1), 91-96 (2016-04-14)
CPA4 belongs to a member of the metallocarboxypeptidase family, and its expression in pancreatic cancer samples and clinical significance are still not investigated until now. In this study, we aimed to evaluate the level of CPA4 in pancreatic cancer samples
Tomohiko Kayashima et al.
Human genetics, 112(3), 220-226 (2003-01-29)
By a search for novel human imprinted genes in the vicinity of the imprinted gene MEST, at chromosome 7q32, we identified the carboxypeptidase A4 gene ( CPA4) in a gene cluster of the carboxypeptidase family, 200 kb centromeric to MEST.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service