Skip to Content
Merck
All Photos(2)

Key Documents

HPA011891

Sigma-Aldrich

Anti-NAV2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Helicase APC down-regulated 1, Anti-Neuron navigator 2, Anti-Pore membrane and/or filament-interacting-like protein 2, Anti-Retinoic acid inducible in neuroblastoma 1, Anti-Steerin-2, Anti-Unc-53 homolog 2, Anti-unc53H2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

LREPSKTALGSSLPGLVNQTDKEKGISSDNESVASCNSVKVNPAAQPVSSPAQTSLQPGAKYPDVASPTLRRLFGGKPTKQVPIATAENMKNSVVISNPHATMTQQGNLDSPSGSGVLSSGSSSPLYSKNVDLNQSPL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NAV2(89797)

Immunogen

Neuron navigator 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NAV2 (Neuron navigator 2) is a 261kDa novel protein of neuron navigator gene family. It consists of several domains such as calponin homology (CH) domain, four coiled-coil (CC) domains, a cytoskeletal interacting region (CSID), and an AAA-domain. It is expressed in the human developing brain and in kidney to lesser extent. It is mainly involved in the neuronal axon guidance and neurite outgrowth in human neuroblastoma cells. Its up-regulated expression has been reported in the colorectal carcinogenesis and thus may have therapeutic implications in this disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71517

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideyuki Ishiguro et al.
Oncogene, 21(41), 6387-6394 (2002-09-06)
To investigate the mechanisms of colorectal carcinogenesis, we searched for genes regulated by adenomatous polyposis coli gene product (APC) and identified a novel gene, termed HELAD1 (helicase, APC down-regulated 1). A recombinant polypeptide representing the ATPases associated with cellular activities
Tamara Maes et al.
Genomics, 80(1), 21-30 (2002-06-25)
We have cloned the gene neuron navigator-1 (NAV1), a human homolog of unc-53, a gene involved in axon guidance in Caenorhabditis elegans. Duplications during evolution gave rise to three human homologs located on chromosomes 1q32.1, 11p15.1, and 12q21.1. NAV1 and
Mark A Marzinke et al.
Archives of biochemistry and biophysics, 540(1-2), 94-100 (2013-10-29)
Neuron navigator 2 (NAV2) is required for all-trans retinoic acid (atRA) to induce neurite outgrowth in human neuroblastoma cells. Further, ectopic overexpression of full-length human NAV2 rescues an axonal elongation defect in the Caenorhabditis elegans unc-53 (NAV2 ortholog) mutant. Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service