Skip to Content
Merck
All Photos(1)

Key Documents

DWK898862802

DURAN® PURE Screw Cap

new

PFA cap, GL 45 thread, with lip seal

Synonym(s):

898862802, DURE, GL45, PURE, cap, closure, lid, packaging, stopper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121821

material

PFA cap
transparent cap

Joint

joint threaded cap

feature

autoclavable

packaging

pack of 5 ea

manufacturer/tradename

DWK Life Sciences DURAN® 898862802

cap diam. × H

53 mm × 27 mm

fitting

GL 45 thread

Looking for similar products? Visit Product Comparison Guide

General description

High-quality plastic screw cap with GL 45 thread size, developed for the needs of the pharmaceutical industry. The DURAN® PURE Lip Seal Cap is manufactured in clean rooms and made from PFA with lip seal design. Designed for use with DURAN® PURE GL 45 Bottles. The screw cap is compatible with autoclaving and other common sterilization methods. DWK Life Sciences provides CoAs with each delivery, assures 100% traceability and change control management. The DURAN® PURE Lip Seal Cap is available with the documentation required by regulatory authorities and meets the requirements of the international pharmacopeias.

Application

storage, pharmaceutical packaging, closure, seal

Features and Benefits

  • PFA
  • High cleanliness due to manufacturing in clean rooms
  • Certificate of Analysis
  • Visual and dimensional inspection
  • 100% traceability of all manufacturing steps
  • Extractables report available
  • A Type III Packaging Material DMF (#28044) for these products has been filed with the FDA. China Center for Drug Evaluation CDE of NMPA registration number B20220000817
  • USP 88 Class VI Pharmaceutical grade materials

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service