DWK818667303
DURAN® PURE Bottle

capacity 5000 mL, amber, graduated, GL 80 thread, with dust cover, without cap, borosilicate glass 3.3 bottle
Synonym(s):
818667303, DURAN PURE bottle, container, pharmaceutical bottle, storage bottle, vessel
About This Item
material
amber
borosilicate glass 3.3 bottle
Joint
joint threaded neck
sterility
non-sterile
feature
autoclavable
graduated
without cap
with dust cover
packaging
pack of 1 ea
manufacturer/tradename
DWK Life Sciences DURAN® 818667303
bottle diam. × H
182 mm × 310 mm
capacity
5000 mL
fitting
GL 80 thread
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Protective cover prevents contamination inside the bottle during storage and transport
- Manufactured from borosilicate glass 3.3
- Certificate of Analysis (CoA)
- 100% visual inspection
- Analysis of hydrolytic resistance according to EP, USP and JP for each batch
- 100% traceability of all manufacturing steps
- A Type III Packaging Material DMF (#28042) for these products has been filed with the FDA
- China Center for Drug Evaluation CDE NMPA registration number B20220000306
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service