Skip to Content
Merck
All Photos(1)

Key Documents

DWK292481456

DURAN® PURE Silicone Seal

new

PTFE-coated seal, platinum cured silicone, GL 25 seal, case of 10 ea

Synonym(s):

DURAN PURE, GL25, gasket, liner, ring, seal, silicone, stopper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121821

material

PTFE-coated seal
gray

packaging

case of 10 ea

manufacturer/tradename

DWK Life Sciences DURAN® 292481456

seal diam. × H

23.5 mm × 3.1 mm

fitting

GL 25 seal

Looking for similar products? Visit Product Comparison Guide

General description

High-quality, PTFE coated silicone seal for use with DURAN® PURE GL 25 Screw Cap

Application

storage, sealing, sample collection

Features and Benefits

  • Certificate of Analysis
  • Manufactured from PTFE coated, platinum cured silicone
  • Packaged in clean room
  • Visual and dimensional inspection
  • USP 88 Class VI Pharmaceutical grade materials

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service