Key Documents
DBS-FRhL-2
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
About This Item
UNSPSC Code:
41106514
biological source
rhesus monkey lung
packaging
tube of 5 μg 89101303-DNA-5UG
pkg of vial of cells 89101303-1VL
growth mode
Adherent
karyotype
2n = 42, diploid no structural abnormalities observed
morphology
Fibroblast
receptors
Not specified
Cell Line Origin
Monkey Rhesus lung
Cell Line Description
A diploid cell line derived from normal male Rhesus monkey fetus. Susceptible to poliovirus types 1-3, Coxsackie A9, parainfluenza 3, rubella and is non-tumourigenic in animals.
Culture Medium
EMEM (EBSS) (M2279) + 2 mM L-Glutamine (G7513) + 1% NEAA (M7145) + 10% FBS / FCS (F2442).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:3, i.e., seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37 °C. Finite lifespan. Cell line has been shown to undergo 51 passages or 74 population doublings.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Legal Information
This material is cited in a US and/or other Patent and may not be used to infringe parent claims. US Patent No. 4,040,905.
Choose from one of the most recent versions:
Certificates of Analysis (COA)