Ma-Mel-27
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 13012485, human skin, Epithelial
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
human skin
description
Human melanoma, metastasis
packaging
tube of 5 μg 13012485-DNA-5UG
pkg of vial of cells 13012485-1VL
growth mode
Adherent
morphology
Epithelial
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Description
Ma-Mel-27 (ESTDAB-168) was established from the cutaneous/subcutaneous metastasis in a 58 year-old female patient with metastatic acrolentiginous melanoma (stage IV). The Ma-Mel-27 cell line is one of more than 170 characterized melanoma cell lines collected during the ESTDAB project (European Searchable Tumour Line Database) part of the European Commissions fifth framework infrastructures programme (contract no. QLRI-CT-2001-01325). ESTDAB contains information on HLA genotype and surface expression of HLA molecules, expression of tumour antigens, antigen processing capability, production of and response to cytokines and chemokines, apoptosis regulation and expression of adhesion/accessory molecules. Such information allows researchers to identify cells possessing specific parameters important for studies of immunity, immunogenetics, gene expression, metastasis, response to chemotherapy, and other tumour biological experimentation.
DNA Profile
STR-PCR Data:
Amelogenin: X
CSF1PO: 10,13
D13S317: 8,11
D16S539: 11,14
D5S818: 12
D7S820: 10,12
THO1: 9.3
TPOX: 8,11
vWA: 17,18
Amelogenin: X
CSF1PO: 10,13
D13S317: 8,11
D16S539: 11,14
D5S818: 12
D7S820: 10,12
THO1: 9.3
TPOX: 8,11
vWA: 17,18
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Fetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) seed new cultures at 1-3 x10,000 cells/cm² using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37°C. Saturation density approx 6 x10,000 cells/cm² .
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service