Skip to Content
Merck
All Photos(2)

Key Documents

AV46899

Sigma-Aldrich

Anti-NPTN antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DKFZp686L2477, Anti-GP55, Anti-GP65, Anti-MGC102805, Anti-Neuroplastin, Anti-Np55, Anti-SDFR1, Anti-SDR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

human, rabbit, horse, dog, rat, bovine, guinea pig, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPTN(27020)

Immunogen

Synthetic peptide directed towards the middle region of human NPTN

Application

Anti-NPTN antibody produced in rabbit is suitable for western blotting at a concentration of 5μg/mL. It is also useful for immunohistochemistry at a concentration of 4-8μg/mL.

Biochem/physiol Actions

NPTN (neuroplastin) gene encodes a single-pass type I membrane protein that belongs to immunoglobulin superfamily and is expressed in brain cortex and cerebellum. It plays a crucial role in the development and maintenance of normal synaptic connections in the cerebellum.

Sequence

Synthetic peptide located within the following region: MEYRINKPRAEDSGEYHCVYHFVSAPKANATIEVKAAPDITGHKRSENKN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hassan Marzban et al.
The Journal of comparative neurology, 462(3), 286-301 (2003-06-10)
Neuroplastin (np) 55 and 65 are immunoglobulin superfamily members that arise by alternative splicing of the same gene and have been implicated in long-term activity-dependent synaptic plasticity. Both biochemical and immunocytochemical data suggest that np55 is the predominant isoform (>95%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service