38548
1-[2-(Maleimido)ethyl]-4-[2-(3,4-dihydro-2H-1-benzopyran-6-yl)-5-oxazolyl]pyridinium triflate
suitable for fluorescence, BioReagent, ≥90% (HPCE)
About This Item
Recommended Products
product line
BioReagent
Quality Level
Assay
≥90% (HPCE)
fluorescence
λex 389 nm; λem 528 nm in 0.1 M phosphate pH 7.0
suitability
suitable for fluorescence
storage temp.
2-8°C
SMILES string
[O-]S(=O)(=O)C(F)(F)F.O=C1C=CC(=O)N1CC[n+]2ccc(cc2)-c3cnc(o3)-c4ccc5OCCCc5c4
InChI
1S/C23H20N3O4.CHF3O3S/c27-21-5-6-22(28)26(21)12-11-25-9-7-16(8-10-25)20-15-24-23(30-20)18-3-4-19-17(14-18)2-1-13-29-19;2-1(3,4)8(5,6)7/h3-10,14-15H,1-2,11-13H2;(H,5,6,7)/q+1;/p-1
InChI key
FGUKXLKALCIXHR-UHFFFAOYSA-M
General description
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service