15613
Hexane, mixture of isomers
≥85% isomers basis (GC), puriss
Synonym(s):
Hexanes, Isohexane
About This Item
Product Name
Hexane, mixture of isomers, puriss., ≥85% isomers basis (GC)
vapor density
3 (vs air)
grade
puriss.
Assay
≥85% isomers basis (GC)
autoignition temp.
437 °F
expl. lim.
7.5 %
technique(s)
GC/GC: suitable
impurities
≤0.002% free acid (as CH3COOH)
≤0.002% non-volatile matter
≤0.005% S-compounds (as S)
≤0.01% aromatic substances (as C6H6) (GC)
≤0.01% water (Karl Fischer)
refractive index
n20/D 1.379 (lit.)
bp
68-70 °C (lit.)
mp
−95 °C (lit.)
density
0.672 g/mL at 25 °C (lit.)
InChI
1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3
InChI key
AFABGHUZZDYHJO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
-9.4 °F
Flash Point(C)
-23 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service