Skip to Content
Merck
All Photos(1)

Key Documents

SMB01401

Sigma-Aldrich

Hydroxycotinine

new

≥95%

Synonym(s):

(3′R,5′S)-3′-Hydroxycotinine, (3S-trans)-3-Hydroxycotinine, trans-3-Hydroxycotinine, Alkaloids, Metabolites, Nicotine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O2
CAS Number:
Molecular Weight:
192.21
MDL number:
UNSPSC Code:
12352200

biological source

synthetic

Quality Level

Assay

≥95%

form

powder or crystals

color

white to light off-white

mp

108-110 °C

solubility

acetonitrile: soluble
chloroform: soluble
methanol: soluble

application(s)

cell analysis
clinical research
general analytical
life science and biopharma
metabolomics

storage temp.

2-8°C

SMILES string

CN1[C@@H](C[C@@H](O)C1=O)C=2C=CC=NC2

InChI

InChI=1S/C10H12N2O2/c1-12-8(5-9(13)10(12)14)7-3-2-4-11-6-7/h2-4,6,8-9,13H,5H2,1H3/t8-,9+/m0/s1

InChI key

XOKCJXZZNAUIQN-DTWKUNHWSA-N

General description

Hydroxycotinine, also known as trans-3′-Hydroxycotinine, is an alkaloid and a cotinine derivative with a hydroxy group substitution at the C-3 position (the 3R,5S-diastereomer). It is functionally related to (-)-cotinine. Hydroxycotinine is classified as an N-alkylpyrrolidine, a member of the pyridines, a pyrrolidine alkaloid, and a member of the pyrrolidin-2-ones. It is a product of CYP2A6 metabolism of the primary nicotine metabolite, cotinine, and serves as a biomarker of nicotine metabolism, holding significant importance in various scientific research applications.

Application

Hydroxycotinine finds application in compound screening libraries, metabolomics, biochemical, phytochemical, and drug discovery research.

Features and Benefits

Versatile and adaptable for wide variety of research and metabolomics applications.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service