Skip to Content
Merck
All Photos(1)

Key Documents

43784

Sigma-Aldrich

Triisobutylphosphine

≥90% (GC)

Synonym(s):

Cytop® 341, Tris(2-methylpropyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]3P
CAS Number:
Molecular Weight:
202.32
Beilstein:
1738664
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

≥90% (GC)

reaction suitability

reagent type: ligand

refractive index

n20/D 1.451 (lit.)

bp

126 °C/50 mmHg (lit.)

density

0.811 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CC(C)CP(CC(C)C)CC(C)C

InChI

1S/C12H27P/c1-10(2)7-13(8-11(3)4)9-12(5)6/h10-12H,7-9H2,1-6H3

InChI key

DAGQYUCAQQEEJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Cytop is a registered trademark of Cytec Technology Corp.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service