Skip to Content
Merck
All Photos(3)

Key Documents

MABS2324

Sigma-Aldrich

Anti-Folic Acid Antibody, clone 1E4.4.7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E4.4.7, monoclonal

mol wt

observed mol wt ~66 kDa

purified by

using protein G

species reactivity (predicted by homology)

all

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

storage temp.

2-8°C

Specificity

Clone 1E4.4.7 is a mouse monoclonal antibody that detects Folic Acid.

Immunogen

KLH-conjugated Folic Acid.

Application

Quality Control Testing

Evaluated by Western Blotting with Folic acid conjugated to BSA.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Folic Acid by Western Blotting.

Tested Applications

ELISA Analysis: A representative lot of this antibody detected Folic Acid in ELISA application.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Folic acid, also known as folate or vitamin B9 plays a crucial role in nucleotide synthesis, and red blood cells. It is an essential nutrient for the early growth and development of the fetal neural tube, potentially reducing the risk of neural tube defects in newborns. In addition to its role in pregnancy, folic acid is important for overall cell growth and metabolism, and it may help reduce the risk of certain birth defects and support heart health. Deficiencies can lead to anemia, poor growth, and various other health issues.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide.

Reconstitution

1 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service