Skip to Content
Merck
All Photos(3)

Key Documents

MABC1607

Sigma-Aldrich

Anti-MMP-11 (Stromelysin-3) Antibody, clone 5ST-4A9

clone 5ST-4A9, from mouse

Synonym(s):

Stromelysin-3, SL-3, ST3, MMP11, Matrixmetalloproteinase 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5ST-4A9, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1λ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MMP11(4320)

General description

Stromelysin-3 (UniProt: P24347; also known as SL-3, ST3, Matrix metalloproteinase-11, MMP-11) is encoded by the MMP11 (also known as STMY3) gene (Gene ID: 4320) in human. MMP-11 is shown to be differentially expressed in tumor tissue and plays a role in tumor invasion. It is shown to be well expressed in stromal cells of breast carcinomas. It has little expression in adult organs under normal physiological conditions. During development, it is expressed in many processes involved in cell migration and tissue morphogenesis. MMP-11 has multiple zinc and calcium binding sites. It binds 2 zinc ions per subunit and one calcium ion per subunit. MMP-11 contains 4 hemopexin domains (aa 291-339; 340-382; 384-432; and 433-480). It is synthesized with a signal peptide (aa 1-31) and a propeptide region (aa 32-97) that are cleaved to generate mature enzyme. It contains a conserved cysteine that is present in the cysteine-switch motif (aa 78-85) that binds the catalytic zinc ion, thus inhibiting the enzyme. The dissociation of the cysteine from the zinc ion activates the enzyme. The precursor is cleaved by a furin endopeptidase. Expression levels of MMP-11 are a reliable indicator of greatest risk for cancer recurrence in breast carcinoma, pancreatic tumors, and colon cancer. (Ref.: Roscilli, G et al. (2014). J. Transl. Med. 12; 54).

Specificity

Clone 5ST-4A9 detects MMP-11 (stromelysin 3) in human cells.

Immunogen

His-tagged full length recombinant human MMP-11 (Stromelysin-3).

Application

Anti-MMP-11 (Stromelysin-3), clone 5ST-4A9, Cat. No. MABC1607, is a mouse monoclonal antibody that detects Stromelysin-3 in human cells and has been tested for use in Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected MMP-11 (Stromelysin-3) in Immunohistochemistry applications in (Ahmad, A., et. al. (1998) Am J Pathol. 152(3):721-8; Nakopoulou, L., et. al. (2002). Mod Pathol. 15(11):1154-61).

Western Blotting Analysis: A representative lot detected MMP-11 (Stromelysin-3) in Western Blotting applications in (von Marschall, Z., et. al. (1998) Gut. 43(5):692-8; Motrescu, E.R., et. al. (2008). Oncogene. 27(49):6347-55; Luo, D., et. al. (1999). J Biol Chem. 274(52):37177-85).

Immunohistochemistry Analysis: A representative lot detected MMP-11 (Stromelysin-3) in Immunohistochemistry applications in (Cribier, B., et. al. (2002). J Am Acad Dermatol. 46(3):408-13; von Marschall, Z., et. al. (1998) Gut. 43(5):692-8; Andarawewa, K.L., et. al. (2005). Cancer Res. 65(23):10862-71; Motrescu, E.R., et. al. (2008). Oncogene. 27(49):6347-55.

Immunoprecipitation Analysis: A representative lot detected MMP-11 (Stromelysin-3) in Immunoprecipitation applications in (Schonbeck, U., et. al. (1999). J Exp Med. 189(5):843-53).

Immunohistochemistry Analysis: A 1:50, 250, 1,000 dilution from a representative detected MMP-11 (Stromelysin-3) in human placenta and human breast cancer tissue.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in PC-3 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected MMP-11 (Stromelysin-3) in 10 µg of PC-3 cell lysate.

Target description

~55 kDa observed; 54.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service