Skip to Content
Merck
All Photos(1)

Key Documents

5248-10X100ML

Millipore

Modified Scholten Semisolid Agar (MSA)

ready-to-use, box of 10 × 125 ± 3 mL, selective for bacteria: Streptococcus spp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217

Agency

ISO 10705-2
ISO 2000

form

box (Semi Solid Media Filled)

feature

ready-to-use

packaging

box of 10 × 125 ± 3 mL

technique(s)

microbiological culture: suitable

pH

7.4 ± 0.2 (25 °C)

application(s)

food and beverages
water monitoring

compatibility

for use with VIRAPREP Kit (AR00206)

storage temp.

2-8°C

suitability

selective for bacteria: Streptococcus spp

General description

Modified Scholten Semisolid Agar (MSA) is a selective and differential microbial culture media. This ready-to-use media comes in a semi-solid form as described in ISO 10705-2: 2000.
Mannitol Salt Agar (MSA) contains higher amount of salt in the composition which inhibits the growth of non-halophilic (salt loving) microbes. Mannitol acts as the energy source, while phenol red serves as a pH indicator, changing color to confirm the presence of target microorganisms.

Application

Modified Scholten Semisolid Agar (MSA) is used to enumeration of somatic coliphages from water, sediment and sludge samples.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service