Skip to Content
Merck
All Photos(1)

Key Documents

1.46656

Millipore

Purified Water

bottle of, ready-to-use, bottle volume 800 mL , filling volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type new cap with 3 septa
ready-to-use

packaging

bottle of

bottle capacity

1000 mL

bottle volume

800 mL , filling volume

impurities

Endotoxins, none detected
Heavy Metals (Pb), none detected
<100 cfu/mL Bacteria

pH

5.0 -7.0

conductivity

<1.1 μS/cm at 20 °C
<1.3 μS/cm at 25 °C

application(s)

cosmetics
food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

General description

Purified Water is a universal solvent derived from non-portable water source which meets the regulatory specifications.
The low microbial counts and pH, synergistically support the usage of purified water in pharma and food industries.

Application

Purified Water is mainly used for microbial media preparations and bulk material for non-parenteral formulations.
Apart from above, purified water finds its application in:
  • Cleaning of non-parenteral production equipment′s.
  • Input for WFI production.
  • Non-sterile tests and assays.

Preparation of Bulk Chemicals.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service