Skip to Content
Merck
All Photos(2)

Key Documents

900604C

Avanti

18:1 Photoclick cholesterol II

Avanti Research - A Croda Brand

Synonym(s):

Photoclick cholesterol II oleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H76N2O2
Molecular Weight:
689.11
UNSPSC Code:
12352211
NACRES:
NA.28

Assay

>99% (TLC)

form

chloroform solution

packaging

pkg of 1x100 μg (900604C-100UG)
pkg of 1x250 μg (900604C-250UG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

100 μg/mL in chloroform
250 μg/mL in chloroform

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@]12[C@](C3(N=N3)CC4C2CC[C@@]5(C)C4CC[C@@H]5[C@@H](CCCC(C)C#C)C)([H])C[C@@H](OC(CCCCCCC/C=C\CCCCCCCC)=O)CC1

InChI

1S/C46H76N2O2/c1-7-9-10-11-12-13-14-15-16-17-18-19-20-21-22-26-43(49)50-37-29-31-45(6)41-30-32-44(5)39(36(4)25-23-24-35(3)8-2)27-28-40(44)38(41)34-46(47-48-46)42(45)33-37/h2,15-16,35-42H,7,9-14,17-34H2,1,3-6H3/b16-15-/t35?,36-,37+,38?,39-,40?,41?,42+,44-,

InChI key

YGTVVJBRLTYRSZ-GMOVSAPTSA-N

Packaging

5 mL Amber Glass Screw Cap Vial(900604C-100UG)
5 mL Amber Glass Screw Cap Vial(900604C-250UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service