Skip to Content
Merck
All Photos(1)

Key Documents

870454O

Avanti

2-AG-d11

Avanti Research - A Croda Brand 870454O

Synonym(s):

2-arachidonoyl glycerol-d11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27D11O4
CAS Number:
Molecular Weight:
389.61
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mg (with screw cap/argon/foil bag (870454O-1mg))

manufacturer/tradename

Avanti Research - A Croda Brand 870454O

shipped in

dry ice

storage temp.

−20°C

General description

2-Arachidonoylglycerol (2-AG) is an arachidonic acid-containing monoacylglycerol. It is a potent endogenous ligand for cannabinoid receptors. 2-AG is found in the brain and hematopoietic cells, such as macrophages, lymphocytes and platelets.

Biochem/physiol Actions

2-Arachidonoylglycerol (2-AG) functions as an agonist of cannabinoid receptors. It weakens synaptic transmission and acts as a neuromodulator. It has cytoprotective activity. 2-AG stimulates the activation of p42/44 MAPK (mitogen-activated protein kinase), p38 MAPK and c-Jun N-terminal kinase.

Packaging

5 mL PTFE Vial with Screw Cap/Argon/Foil Bag (870454O-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Endocannabinoid 2-Arachidonoylglycerol Regulates Platelet Function.
Baldassarri S, et al.
Blood (2006)
The Endocannabinoid 2-Arachidonoylglycerol Regulates Platelet Function.
Baldassarri S, et al.
Blood (2006)
2-Arachidonoylglycerol, an endogenous cannabinoid receptor ligand, induces rapid actin polymerization in HL-60 cells differentiated into macrophage-like cells
Gokoh M, et al.
The Biochemical Journal, 386(3) (2005)
Evidence That the Cannabinoid CB1 Receptor Is a 2-Arachidonoylglycerol Receptor Structure-Activity Relationship of 2-Arachidonoylglycerol, Ether-linked Analogues, and Related Compounds
Sugiura T, et al.
The Journal of Biological Chemistry, 274(5) (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service