Skip to Content
Merck
All Photos(1)

Key Documents

870305P

Avanti

16:0 Ptd Ethylene Glycol

Avanti Research - A Croda Brand 870305P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phospho(ethylene glycol) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H72O9PNa
CAS Number:
Molecular Weight:
714.93
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (870305P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870305P

lipid type

headgroup modified lipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCO)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C37H73O9P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(39)43-33-35(34-45-47(41,42)44-32-31-38)46-37(40)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h35,38H,3-34H2,1-2H3,(H,41,42);/q;+1/p-1/t35-;/m1./s1

InChI key

YDTNKUACKJHTLJ-RUQJKXHKSA-M

Application

16:0 Ptd Ethylene Glycol or 1,2-dipalmitoyl-sn-glycero-3-phospho(ethylene glycol), is suitable for synthesis of lipid-(succinate)-mPEG conjugate, which is used in lipid-polymer hybrid nanoparticles preparation.

Packaging

5 mL Amber Glass Screw Cap Vial (870305P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service