Skip to Content
Merck
All Photos(1)

Key Documents

860904P

Avanti

19:0-12:0-19:0 D5 TG

Avanti Research - A Croda Brand 860904P, powder

Synonym(s):

1,3(d5)-dinonadecanoyl-2-dodecanoyl-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H97D5O6
CAS Number:
Molecular Weight:
840.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860904P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860904P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C(C([2H])([2H])OC(CCCCCCCCCCCCCCCCCC)=O)(OC(CCCCCCCCCCC)=O)C([2H])([2H])OC(CCCCCCCCCCCCCCCCCC)=O

InChI

1S/C53H102O6/c1-4-7-10-13-16-19-21-23-25-27-29-31-34-36-39-42-45-51(54)57-48-50(59-53(56)47-44-41-38-33-18-15-12-9-6-3)49-58-52(55)46-43-40-37-35-32-30-28-26-24-22-20-17-14-11-8-5-2/h50H,4-49H2,1-3H3/i48D2,49D2,50D

InChI key

YVHBSDKZPZMEEF-BXJPYAOGSA-N

Application

19:0-12:0-19:0 D5 TG is suitable for use as a standard to identify lipid metabolites by ultra-performance liquid chromatography - tandem mass spectrometer (UPLC-MS).

Packaging

5 mL Amber Glass Screw Cap Vial (860904P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Effects of Age and Reproduction on the Lipidome of Caenorhabditis elegans
Wan QL, et al.
Oxidative Medicine and Cellular Longevity, 2019 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service