Skip to Content
Merck
All Photos(1)

Key Documents

860158P

Avanti

C18:0 GM1(Neu5Gc) Ceramide (d18:1/18:0)

Avanti Research - A Croda Brand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C73H131N3O32
CAS Number:
Molecular Weight:
1562.82
UNSPSC Code:
12352211
NACRES:
NA.79

biological source

synthetic

Assay

>99 (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

Application

Lipids research

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service