Z743851
ART® Non-Filtered Tip
HLT 1000G, size 1000 μL, racked, sterile
Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
About This Item
racked
Recommended Products
sterility
sterile
feature
HLT 1000G
racked
packaging
pkg of 4X800 ea
manufacturer/tradename
Molecular BioProducts 3592
technique(s)
PCR: suitable
size
1000 μL
volume
1000 μL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Complete selection of non-filtered and specialty tips provides ART quality, performance, and reliability.
- Racks allow saving time by offering quick and easy access to pipette tips.
- The front latch closes and locks to provide safe and secure storage
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service