Skip to Content
Merck
All Photos(1)

Key Documents

W336900

Sigma-Aldrich

Isobutyl isovalerate

natural (US), ≥99%, FG

Synonym(s):

2-Methylpropyl 3-methylbutyrate, Isobutyl 3-methyl butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O2
CAS Number:
Molecular Weight:
158.24
FEMA Number:
3369
EC Number:
Council of Europe no.:
568
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.472
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural (US)
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural (US)

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥99%

refractive index

n20/D 1.4060

density

0.851 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; green; fruity; sweet

SMILES string

CC(C)COC(=O)CC(C)C

InChI

1S/C9H18O2/c1-7(2)5-9(10)11-6-8(3)4/h7-8H,5-6H2,1-4H3

InChI key

KEBDNKNVCHQIJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service