GF46470304
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3
1000mm tube, 20.0mm outside diameter, 1.5mm wall thickness, 17mm inside diameter, annealed
Synonym(s):
FE247800
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11171600
NACRES:
NA.23
Recommended Products
form
tubes
manufacturer/tradename
Goodfellow 464-703-04
L × wall thickness
1000 mm × 1.5 mm
O.D. × I.D.
20.0 mm × 17 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service