Skip to Content
Merck
All Photos(2)

Key Documents

GF36721762

Platinum

foil, not light tested, 100x100mm, thickness 0.0025mm, as rolled, 99.95%

Synonym(s):

Platinum, PT000206

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 367-217-62

resistivity

10.6 μΩ-cm, 20°C

size × thickness

100x100 mm × 0.0025 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayako Tokuda
Kokubyo Gakkai zasshi. The Journal of the Stomatological Society, Japan, 71(1), 18-26 (2004-04-24)
To fabricate a prosthesis which is functionally harmonized with the stomatognathic system, the interocclusal relationship must be reproduced as accurately as possible. This study investigated the influence of the occlusal contacts upon tooth displacement and clarified the optimal occlusal contacts
Martin Schuettler et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2008, 3212-3215 (2009-01-24)
Standard interconnection technologies are reviewed in respect to their applicability to electrically and mechanically connect laser-patterned nerve electrodes made from silicone rubber and platinum foil to wires and screen printed alumina substrates. Laser welding, gap welding, microflex ball bonding, and
Yo Shibata et al.
The International journal of oral & maxillofacial implants, 19(2), 177-183 (2004-04-23)
This study evaluated the quality of coatings on titanium surfaces prepared by discharging in Hanks' balanced salt solution without organic molecules (HBSS-). 10 x 10 x 1.0-mm titanium plates were used as cathodes of a coating device developed in the
J A Sorensen et al.
The Journal of prosthetic dentistry, 67(1), 16-22 (1992-01-01)
This study evaluated the marginal fidelity and microleakage of porcelain veneers made with the platinum foil and refractory die techniques. Maxillary incisors, matched for size and amount of enamel, were prepared with 0.5 mm uniform intraenamel reduction. The indirectly made
Ayumi Ikeda et al.
Journal of medical and dental sciences, 52(4), 195-202 (2006-05-04)
The purpose of this study was to clarify the influence of changes of occlusal contacts on adjacent tooth displacement during articulation. The amount and direction of displacement of the maxillary left first molar and adjacent teeth were measured using a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service