Skip to Content
Merck
All Photos(2)

Key Documents

DWKWGF230P

Azlon Square Shouldered Wash Bottles with jet-tip aperture

low-density polyethylene bottle, capacity 250 mL

Synonym(s):

WGF230P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

material

low-density polyethylene bottle

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGF230P

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for solvent and water dispensing within the laboratory

Features and Benefits

Precision jet tip for fine control

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service