DWKBGG312
Azlon Specimen Container With hinged cap
low-density polyethylene container, capacity 25 mL
Synonym(s):
BGG312
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NB.43
Recommended Products
material
low-density polyethylene container
packaging
pkg of 10 ea
manufacturer/tradename
Azlon BGG312
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Suitable for generall sample collection and storage applications
Features and Benefits
Integral hinged cap
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service