Skip to Content
Merck
All Photos(2)

Key Documents

DWKBGG306

Azlon Specimen Container With hinged cap

low-density polyethylene container, capacity 5 mL

Synonym(s):

BGG306

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NB.43

material

low-density polyethylene container

packaging

pkg of 20 ea

manufacturer/tradename

Azlon BGG306

capacity

5 mL

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for generall sample collection and storage applications

Features and Benefits

Integral hinged cap

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service