DWKBGE530P
Azlon Dispensing Bottles with retractable pivot nozzle
round bottom, low-density polyethylene, capacity 250 mL
Synonym(s):
BGE530P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121818
NACRES:
NB.43
material
low-density polyethylene
round bottom
sterility
non-sterile
packaging
pkg of 10 ea
manufacturer/tradename
Azlon
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Suitable for as dropping bottle and excellent for dispensing gels
Features and Benefits
Pivotting nozzle closure
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service