Skip to Content
Merck
All Photos(2)

Key Documents

DWKBGE520P

Azlon Dispensing Bottles with retractable pivot nozzle

round bottom, low-density polyethylene, capacity 60 mL

Synonym(s):

BGE520P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121818
NACRES:
NB.43

material

low-density polyethylene
round bottom

sterility

non-sterile

packaging

pkg of 10 ea

manufacturer/tradename

Azlon

capacity

60 mL

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for as dropping bottle and excellent for dispensing gels

Features and Benefits

Pivotting nozzle closure

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service