Skip to Content
Merck
All Photos(1)

Key Documents

DWKBDD312P

Azlon Graduated Griffin-Style Low Form Beakers

polymethylpentene, capacity 25 mL

Synonym(s):

BDD312P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121803
NACRES:
NB.43

material

polymethylpentene

packaging

pkg of 12 ea

manufacturer/tradename

Azlon

storage condition

room temperature

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for general laboratory use and management of liquid samples

Features and Benefits

Autoclavable to 150°C

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service