DWK241254404
Duran® Dropping Funnel
pressure tubing with pressure equalisation tube
, standard ground cone 24/29, stopcock and key retaining device, capacity 500 mL
Synonym(s):
241254404
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 241254404
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
Application
uniform and metered liquid supply to a reaction mixture. The rate of supply can be adjusted
Features and Benefits
With pressure equalisation tube, standard ground cone, standard ground stopcock and retaining device; cylindrical, with scale, ground joint and pressure equalisation tube
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service