Skip to Content
Merck
All Photos(2)

Key Documents

DWK231752155

Disposable Culture Tube

Synonym(s):

231752155

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121706
NACRES:
NB.13

material

polypropylene screw cap
soda-lime glass vial

Joint

Joints (GL 18)

feature

seal (rubber)

packaging

pkg of 100 ea

manufacturer/tradename

231752155

W × L

16 mm × 160 mm

Looking for similar products? Visit Product Comparison Guide

Application

Preparation cell culture, sample collecting, storage

Features and Benefits

straight rim, round bottom, not reusable, with screw cap

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service