DWK218663604
DURAN® wide mouth amber laboratory bottle
without cap, bottle capacity (250 mL), round amber glass bottle, non-sterile
Synonym(s):
218663604
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
round amber glass bottle
sterility
non-sterile
feature
without cap
packaging
case of 10 ea
manufacturer/tradename
DURAN® 218663604
bottle capacity
(250 mL)
fitting
GL 80 cap
GL 80 neck
General description
UV protection up to approx. 500 nm due to external colouration. The only external application of the colour ensures that internal surfaces have standard DURAN glass properties. Special thread design for rapid opening and closing in less than a turn Optimum filling and pouring and particularly easy cleaning. Spoons, spatulas and tweezers can be manipulated with ease.
Even large-volume funnels can be inserted into the bottles without any difficulty. Stability due to the large base areaEasy-to-read scale with graduation marks to estimate the contentFired-on, highly durable white ceramic print.
Even large-volume funnels can be inserted into the bottles without any difficulty. Stability due to the large base areaEasy-to-read scale with graduation marks to estimate the contentFired-on, highly durable white ceramic print.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)