DWK212171708
Duran® Super Duty Erlenmeyer Flask
graduated, narrow-mouth neck
Synonym(s):
212171708
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
narrow-mouth neck
feature
graduated
packaging
pkg of 10 ea
manufacturer/tradename
DURAN® 212171708
capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
Application
For demanding applications, Heating Diluting solutions, Mixing several components, Measuring liquid amounts, Application note: To avoid breakages due to thermal stress, uniform and slow heating of SUPER DUTY products is recommended., Boro 3.3
Features and Benefits
Higher impact strength, mechanical strength and stability achieved by reinforced rim; Due to reduced risk of accidental breakage the risk of injuries is minimised and the safety of the user is increased; Economical due to longer life span and outstanding corrosion protection; White easy-to-read scale with graduation marks to estimate the content; Fired-on, highly durable white ceramic
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service