BR733646
BRAND® robotic filter tips

PP, 1 - 20 µl, TipRack, BIO-CERT® LIQUID HANDLING QUALITY, 960EA
Synonym(s):
1-20 ul, 733646, BRAND plates, TipRack, filter tips, microtiter plate, pipette tips, robotic tips, tips, transfer tips
About This Item
PCR clean
RNase and DNase free
endotoxin free
filter
pyrogen free
Recommended Products
material
polypropylene tip
sterility
non-sterile
feature
ATP free
PCR clean
RNase and DNase free
endotoxin free
filter
pyrogen free
packaging
pack of 960 ea
manufacturer/tradename
BRAND 733646
volume
1—20 μL
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- BIO-CERT® LIQUID HANDLING QUALITY: Free of DNA, RNase, DNase, Pyrogens, PCR-Inhibitors and ATP
- For use with the BRAND® Liquid Handling Station pipetting robot
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service