Skip to Content
Merck
All Photos(1)

Key Documents

940917

Chloride-rich argyrodite lithium phosphorus sulfide

new

Li5.5PS4.5Cl1.5 solid electrolyte, powder, battery grade

Synonym(s):

Li-argyrodite, Lithium chloride phosphorotetrathioate sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Li5.5PS4.5Cl1.5
CAS Number:
Molecular Weight:
266.62
UNSPSC Code:
12352302

grade

battery grade

Quality Level

Assay

99.5%

form

powder

mol wt

266.62 g/mol

composition

Li5.5PS4.5Cl1.5

conductivity

4.8 mS/cm (typical ionic conductivity, cold-pressed pellet at room temperature)

density

1.66 g/mL

application(s)

battery manufacturing

SMILES string

[Li].[S].[Cl].S=P([S-])([S-])[S-]

InChI

1S/Cl.Li.H3PS4.S/c;;2-1(3,4)5;/h;;(H3,2,3,4,5);/p-3

General description

Li5.5PS4.5Cl1.5 is a chloride-rich argyrodite sulfide electrolyte that crystallizes in a cubic crystal structure (space group F-43m). It manifests as a light grey powder with a remarkable crystal structure that facilitates high ionic conductivity, typically ranging from 4 to 10 mS/cm at 298 K. This conductivity is nearly four times greater than that of Li6PS5Cl under identical processing conditions. The substantial enhancement in ionic conductivity is attributed to the weakened interactions between the mobile Li-ions and the surrounding framework anions, resulting from the substitution of divalent S2- by monovalent Cl-. Additionally, increased site disorder and a higher lithium vacancy population contribute to the improved ionic conductivity.

Application

Li5.5PS4.5Cl1.5 is primarily employed as a catholyte in solid-state batteries, often in conjunction with LiNbO3-coated NMC (LiNixMnyCozO2) or LFP (LiFePO4) cathodes. It exhibits compatibility with silicon and graphite anodes but is not directly compatible with lithium metal anodes. The material′s high ionic conductivity and low activation barrier (0.29 eV) render it an ideal candidate for room-temperature solid-state batteries, offering a safer and more stable alternative to traditional organic electrolytes. By virtue of its crystal structure and conductivity up to 9 mS/cm at 298 K, Li5.5PS4.5Cl1.5 can significantly enhance the performance of solid-state batteries, providing excellent energy storage capabilities and improved safety features. The material′s ability to exhibit high ionic conductivity under various processing conditions makes it a valuable component in the development of advanced solid-state battery technologies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service