Skip to Content
Merck
All Photos(1)

Key Documents

940240

Sigma-Aldrich

Ketjenblack® EC-600JD

new

conductive carbon black

Synonym(s):

Carbon black

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
UNSPSC Code:
12171603
EC Index Number:
215-609-9

grade

battery grade

Quality Level

description

surface area ~1400 m2/g (BET)

form

solid

pH

9.0-10.5

bp

4200 °C (lit.)

mp

3550 °C (lit.)

bulk density

0.100‑0.120 g/cm3

application(s)

battery manufacturing

SMILES string

[C]

General description

Ketjenblack EC-600JD is a highly pure carbon black with a distinctive mesoporous structure. As a result, it offers a high surface area of around 1400 m2/g (BET) and a high electronic conductivity.

Application

Ketjenblack EC-600JD is widely used in numerous antistatic and conductive purposes for research and development. Commonly, Ketjenblack EC-600JD is used as a support for electrocatalysts in the electrodes of fuel cells. The high surface area of Ketjenblack EC-600JD, along with its capability to create conductive networks, makes it a great choice for supporting nanoparticle catalysts. Ketjenblack serves as a flexible base for creating catalyst composites: for example, catalyst nanoparticles can be dispersed and deposited onto the Ketjenblack carbon, or the catalyst can be grown directly on the Ketjenblack carbon. This adaptability is boosted by Ketjenblack′s thermal stability, which accommodates various synthesis techniques involving heat treatment or annealing even to 1000 °C without affecting the material′s integrity or conductivity. This thermal resilience is particularly useful in oxides, nitrides, and ceramic catalysts, which often benefit from high-temperature processing. In the production of battery electrodes, especially those incorporating nanoparticle cathode materials or organic cathode materials, Ketjenblack EC-600JD proves beneficial. When used in minor proportions (around 5 wt%), it significantly enhances the electronic conductivity of the electrode film, which boosts battery efficiency. Moreover, Ketjenblack EC-600JD is a widely recognized choice for anti-static enhancements in polymer composites. Its high surface area allows it to establish percolation networks even at low weight fractions (below 2 wt% when adequately dispersed), thereby minimizing changes to the mechanical and rheological properties of the polymer. Dispersing Ketjenblack EC-600JD can present challenges, especially within viscous mediums. For instance, when applied in battery electrodes, it is advisable to thoroughly disperse Ketjenblack EC-600JD into the solvent before adding the PVDF binder or cathode active substances to minimize the formation of carbon clusters in the final product.
battery manufacturing

Legal Information

Ketjenblack is a registered trademark of Nouryon Functional Chemicals B.V. or affiliates

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service